Tisk…
VWR®, Magnetické míchačky, profesionální řada
Dodavatel: VWR Collection
442-1266EA 46080 CZK
442-1266 442-1271 442-1272 442-1267
VWR®, Magnetické míchačky, profesionální řada
Míchadla Michačky magnetické s ohřevem
Designed for applications requiring exceptional accuracy, stability, and repeatability these magnetic hotplate stirrers are equipped with superior heating and mixing capabilities. With temperature ranges up to 500 °C and stirring speeds reaching 1600 rpm, the VWR® magnetic hotplate stirrers are equipped to handle any research, academic and industrial application. Available in two sizes.
  • Excellent temperature uniformity and powerful stirring
  • Easy to read, large LCD displays temperatures, speed and timer settings
  • Cool touch, chemical-resistant housing