Tisk…
Programmable magnetic stirrer, MagStir Genie®
Dodavatel: SCIENTIFIC INDUSTRIES ELMIS
SI-0302
442-0216EA 23320 CZK
442-0216
Programmable magnetic stirrer, MagStir Genie®
Míchadla Míchačky magnetické
Programmable magnetic stirrer with wide range of applications including stirring of cell cultures and viscous solutions.
  • Low profile stirrer with precise and reproducible control
  • Three power levels; low, medium and high - depending on viscosity of the sample
  • Manual mode provides stirring-, reverse direction- and time (1 second to 99 hours) functions
  • Profiles mode allows to program speed, ramp rate, stirring direction, time (including pause), and power level in up to nine distinct steps
  • Automatic mode recalls and executes saved programs
  • Up to four stirring
More Product Information