Tisk…

N,N'-Bis(acryloyl)cystamine ≥97%

Dodavatel: Thermo Fisher Scientific
Varování


Packaged under argon.

44132.03 44132.06 44132.14
44132.03EA 3430 CZK
44132.03 44132.06 44132.14
N,N'-Bis(acryloyl)cystamine ≥97%
N,N'-Bis(acryloyl)cystamine

Purity: 98% CAS No.: 60984-57-8 Molecular Formula: C10H16N2O2S2 Formula Weight: 260.37 Storage Temperature: -30°C to -10°C Physical Form: Powder, Packaged under argon Appearance: White Melting Point: 123-124° MDL No.: MFCD00036225


Applications: Used as a reversible cross-linker for polyacrylamide gels

Vzorec: C₁₀H₁₆N₂O₂S₂
Bod tání: 123…124 °C
Storage Temperature: Freezer
MDL Number: MFCD00036225
CAS číslo: 60984-57-8
EINECS: 262-546-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR