Tisk…

Vanad ≥99.8% (metals basis), wire, Ø 0.075 mm (0.003 in)

Dodavatel: Thermo Fisher Scientific

44108.G1 44108.G5
44108.G1EA 10950 CZK
44108.G1 44108.G5
Vanad ≥99.8% (metals basis), wire, Ø 0.075 mm (0.003 in)
Vanad

Purity: 99.8% CAS No.: 7440-62-2 Physical Form: ≈0.03g/m Odour: Odourless MDL No.: MFCD00011453

Vzorec: V
Bod varu: 3477 °C (1013 hPa)
Bod tání: 1896 °C
Hustota: 6,015 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011453
CAS číslo: 7440-62-2
EINECS: 231-171-1
Merck Index: 12,10053

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR