Tisk…
Stirrers with stirrer bearing
Dodavatel: Lenz Laborglas GmbH & CO.KG
06310000 06310503 06310501 06310505 06310502 06310506 06310504 06310507
441-9711EA 8120 CZK
441-9711 LENZ06310503 441-0383 LENZ06310505 LENZ06310502 441-0385 441-0384 441-0386
Stirrers with stirrer bearing
Míchadla Nástroje na míchání
DURAN®, borosilicate glass, PTFE blade and securing screw. Gas tight closure for vacuum stirring apparatus up to 10⁻³ mbar at continuous operational speed of 800 min⁻¹.
  • Ideal for use with 250 to 2000 ml round bottomed flasks
  • Includes PP screw cap with GL 25 closure