Tisk…
Overhead stirrer shafts
Dodavatel: COWIE
005.440.2 005.450.2 005.455.2 005.465.2 005.240.2 005.250.2 005.255.2 005.265.2 005.340.2 005.350.2 005.355.2 005.365.2 005.355.1.2 005.365.1.2 005.0265.12.2 005.0465.12.2 005.255.10.2 005.430.8.2 005.440.8.2 005.450.8.2 005.455.10.2 005.0465.10.2 005.0475.10.2 005.0475.12.2 005.04100.12.2 005.330.8.2 005.340.8.2 005.350.8.2 005.355.10.2 005.0365.10.2 005.0375.10.2 005.0365.12.2 005.0375.12.2 005.03100.12.2 005.230.8.2 005.240.8.2 005.250.8.2 005.0265.10.2 005.0275.10.2 005.0275.12.2 005.02100.12.2 005.0475.16.2 005.0465.2 005.04100.16.2 005.0375.16.2 005.0375.1.2 005.0365.1.2 005.03100.16.2 005.0275.2 005.230.2 005.02100.16.2 005.430.2 005.0275.16.2 005.0475.2 005.330.2 005.0265.2
441-9521EA 2600 CZK
441-9521 441-9522 441-9523 441-9524 441-9530 441-9532 441-9533 441-9534 441-9571 441-9572 441-9573 441-9574 441-9575 441-9576 441-0435 COWI005.0465.12.2 441-0449 441-0451 441-0452 441-0453 441-0454 441-0443 441-0445 441-0446 441-0441- 441-0377 441-0378 441-0379 441-0380 441-0366 441-0369 441-0367 441-0370 441-0363 441-0373 441-0374 441-0375 441-0434 441-0437 441-0438 441-0432 441-0447 441-0444 441-0442 441-0371 441-0368 441-0365 441-0364 441-0440 441-0372 441-0433 441-0450 441-0439 441-0448 441-0376 441-0436
Overhead stirrer shafts
Míchadla Nástroje na míchání
PTFE, with encapsulated steel reinforced drive shaft. Chemically inert. The smooth surface reduces contamination and makes cleaning easier.
  • Non scratch, prevents agitation vessels from being damaged
  • Temperature resistant from −200 to +280 °C
Upozornení: EX denotes exposed end, these stirrers have a stainless steel shaft core with an exposed end for rigid clamping.