Tisk…

Amberlyst® 15 (H), wet ion exchange resin

Dodavatel: Thermo Fisher Scientific

44079.30 44079.A1 44079.A7
44079.30EA 1430 CZK
44079.30 44079.A1 44079.A7
Amberlyst® 15 (H), wet ion exchange resin
Amberlyst® 15 (H)

CAS No.: 39389-20-3 Molecular Formula: Styrene-DVB; Strongly acidic macroreticular resin Physical Form: Beads Sensitivity: Hygroscopic MDL No.: MFCD00145841

Vzorec: (C₁₀H₁₀·C₈H₈O₃S)x
Storage Temperature: Ambient
MDL Number: MFCD00145841
CAS číslo: 39389-20-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR