Tisk…

Ytterbium(III) bromide hydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Dodavatel: Thermo Fisher Scientific
Varování

44057.09 44057.18
44057.09EA 3540 CZK
44057.09 44057.18
Ytterbium(III) bromide hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Ytterbium(III) bromide hydrate

Purity: 99.99% CAS No.: 15163-03-8 Molecular Formula: YbBr3.xH2O Formula Weight: 412.77(anhy) Physical Form: Lump

Vzorec: Br₃Yb·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00049613
CAS číslo: 15163-03-8
EINECS: 237-350-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR