Tisk…

Sulphur standard dissolved in oil, 5.000 µg/g S in mineral oil Specpure® pro wear metal analysis

Dodavatel: Thermo Fisher Scientific

43870.18
43870.18EA 3470 CZK
43870.18
Sulphur standard dissolved in oil, 5.000 µg/g S in mineral oil Specpure® pro wear metal analysis
Standardy Standardy kationické Standardy - síra

Physical form: liquid
MDL No.: MFCD00242296

Objednat

Produktová specifikace

Environmentally Preferable
MDL čí. MFCD00242296

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR