Tisk…

Hliník ≥99.9995% (metals basis), foil, Puratronic®, annealed, Thickness: 0.2 mm (0.008 in)

Dodavatel: Thermo Fisher Scientific

43777.GH 43777.HP
43777.GHEA 4710 CZK
43777.GH 43777.HP
Hliník ≥99.9995% (metals basis), foil, Puratronic®, annealed, Thickness: 0.2 mm (0.008 in)
Hliník

Purity: 99.9995% CAS No.: 7429-90-5 Physical Form: ≈5.4g/100x100mm, Temper: soft Odour: Odourless MDL No.: MFCD00134029

Vzorec: Al
Bod varu: 2327 °C (1013 hPa)
Bod tání: 660 °C
Hustota: 2,7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS číslo: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR