Tisk…
Crucible, quartz
Dodavatel: Thermo Fisher Scientific
43498.KG
43498.KGEA 6180 CZK
43498.KG
Crucible, quartz
Kelímky
Fused quartz crucible and lid.
  • High chemical purity
  • High corrosion resistance
  • High melting point
  • Extreme hardness and low coefficient of thermal expansion
  • Excellent electrical insulation qualities and optical transmission from ultra-violet to infra-red