Tisk…

Oxid arzenitý ≥99.5% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

43488.30 43488.14 43488.A1
43488.30EA 3140 CZK
43488.30 43488.14 43488.A1
Oxid arzenitý ≥99.5% (metals basis)
Oxid arzenitý

Purity: 99.5% CAS No.: 1327-53-3 Molecular Formula: As2O3 Formula Weight: 197.84 Physical Form: -100 Mesh Powder Density: 3.74 Melting Point: 312.3° Boiling Point: 465° subl. Solubility: Slightly soluble in cold water. Soluble in boiling water, HCl, alkali hydroxide. Insoluble in alcohol, chloroform, and ether Merck Reference: 14,804 UN No.: 1561 MDL No.: MFCD00003433

Vzorec: As₂O₃
Bod varu: 465 °C (subl.)
Hustota: 3,74
Storage Temperature: Ambient
MDL Number: MFCD00003433
CAS číslo: 1327-53-3
EINECS: 215-481-4
UN: 1561
ADR: 6.1,II
Merck Index: 14,804

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR