Tisk…

Tellurium tetrabromide, ultra dry ≥99.999% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Tellurium(IV) bromide

Ampuled under argon

43439.09 43439.18
43439.09EA 6010 CZK
43439.09 43439.18
Tellurium tetrabromide, ultra dry ≥99.999% (metals basis)
Tellurium tetrabromide

Purity: 99.999% CAS No.: 10031-27-3 Molecular Formula: TeBr4 Formula Weight: 447.27 Physical Form: Powder, Ampouled under argon Density: 4.31 Melting Point: 380° Boiling Point: 420° dec. Merck Reference: 14,9125 UN No.: 2923 MDL No.: MFCD00049575

Vzorec: Br₄Te
MW: 447.22
Bod varu: 420 °C (rozkl.)
Bod tání: 380 °C
Hustota: 4.3
Storage Temperature: Ambient
MDL Number: MFCD00049575
CAS číslo: 10031-27-3
EINECS: 233-090-7
UN: 2923
ADR: 8 II
Merck Index: 14,9125

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR