Tisk…

hořčík ≥99.95% (metals basis), slug, annealed, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)

Dodavatel: Thermo Fisher Scientific

43297.09 43297.18
43297.09EA 4570 CZK
43297.09 43297.18
hořčík ≥99.95% (metals basis), slug, annealed, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
hořčík

Purity: 99.95% CAS No.: 7439-95-4 Physical Form: ≈0.09g/slug Odour: Odourless MDL No.: MFCD00085308

Vzorec: Mg
Bod varu: 1107 °C (1013 hPa)
Bod tání: 651 °C
Hustota: 1,74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS číslo: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III
Merck Index: 13,05675

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR