Tisk…

hořčík ≥99.9% (metals basis), foil, hard, Thickness: 0.05 mm (0.002 in)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

43202.FF 43202.FI
43202.FFEA 6760 CZK
43202.FF 43202.FI
hořčík ≥99.9% (metals basis), foil, hard, Thickness: 0.05 mm (0.002 in)
hořčík

Purity: 99.9% CAS No.: 7439-95-4 Physical Form: ≈0.05g/25x25mm, Temper: as rolled Odour: Odourless UN No.: 1869 MDL No.: MFCD00085308


Sacrificial electrode for other material, deoxidizer for Cu

Vzorec: Mg
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS číslo: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR