Tisk…
Homogeniser, Dounce type, KIMBLE®
Dodavatel: DWK Life Sciences
SP885300-0002 SP885300-0007 SP885300-0015 SP885300-0040 SP885300-0100 SP885301-0002 SP885301-0007 SP885301-0015 SP885301-0040 SP885302-0002 SP885302-0007 SP885302-0015 SP885302-0040 SP885303-0002 SP885303-0007 SP885303-0015 SP885303-0040
432-0250EA 20190 CZK
432-0250 432-0251 432-0252 432-0253 SCERSP885300-0100 432-0254 432-0255 432-0256 432-0257 432-0258 432-0259 432-0260 432-0261 432-0262 432-0263 432-0264 432-0265
Homogeniser, Dounce type, KIMBLE®
Homogenizátory Homogenizátory na tkáně
Designed primarily for cellular work where the nucleus remains intact after homogenisation.
  • All-glass construction
  • Two pestles are supplied with each complete unit
  • Large clearance pestle is used for the initial sample reduction
  • Small clearance pestle is used to form the final homogenate
  • Replacement components are available and completely interchangeable