Tisk…
Sterilised filter bags, Whirl-Pak®
Dodavatel: NASCO FORT ATKINSON
B01318WA B01348WA B01416WA B01547WA B01525WA B01488WA
432-0169EA 25220 CZK
432-0169 432-0170 432-0464 432-0471 432-0469 432-0468
Sterilised filter bags, Whirl-Pak®
Sáčky Sáčky na odběr moči
Sterilised post-manufacturing and specially designed for use with homogeniser blenders. HACCP, EPA, USDA, FDA approved for food contact.
  • Plus 1 sterilisation, post-manufacturing sterilisation: Increased sample integrity, perforated closure to maintain sterility
  • 330 micron filter layer collecting, blending, pipetting
  • Convenient write-on strip for easy labelling
  • Puncture-proof closure tabs: Inner and outer tapes for longevity and stability, wire for easy opening, pouring and closing
  • Opening pull tabs: For easy opening, non slip textured Tyvek
  • Continuous
More Product Information