Tisk…

Ammonium hexafluorotitanate ≥99.99% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování

43192.22 43192.14 43192.36
43192.22EA 13460 CZK
43192.22 43192.14 43192.36
Ammonium hexafluorotitanate ≥99.99% (metals basis)
Ammonium hexafluorotitanate

Purity: 99.99% CAS No.: 16962-40-6 Molecular Formula: (NH4)2TiF6 Formula Weight: 197.95 Physical Form: Crystalline Appearance: White MDL No.: MFCD00010888

Vzorec: H₈F₆N₂Ti
Storage Temperature: Ambient
MDL Number: MFCD00010888
CAS číslo: 16962-40-6
EINECS: 241-036-9
UN: 3288
ADR: 6.1,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR