Tisk…

Hexafluorkremičitan amonný ≥99.999% (metals basis), Puratronic®

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Ammonium silicofluoride Ammonium hexafluorosilicate, Ammonium fluorosilicate, Ammomium fluosilicate

43184.09 43184.30 43184.18
43184.09EA 2670 CZK
43184.09 43184.30 43184.18
Hexafluorkremičitan amonný ≥99.999% (metals basis), Puratronic®
Hexafluorkremičitan amonný

Purity: 99.999% CAS No.: 16919-19-0 Molecular Formula: (NH4)2SiF6 Formula Weight: 178.14 Physical Form: Crystalline Melting Point: dec. UN No.: 2854 MDL No.: MFCD00010887

Vzorec: H₈F₆N₂Si
Bod tání: Dec.
Hustota: 2,01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010887
CAS číslo: 16919-19-0
EINECS: 240-968-3
UN: 2854
ADR: 6.1,III
Merck Index: 14,527

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR