Tisk…

Dusičnan hlinitý nonahydrát ≥99.999% (metals basis), Puratronic® low in mercury

Dodavatel: Thermo Fisher Scientific
Nebezpečí

43145.22 43145.14 43145.36
43145.22EA 12840 CZK
43145.22 43145.14 43145.36
Dusičnan hlinitý nonahydrát ≥99.999% (metals basis), Puratronic® low in mercury
Dusičnan hlinitý nonahydrát

Purity: 99.999% CAS No.: 7784-27-2 Molecular Formula: Al(NO3)3.9H2O Formula Weight: 375.13 (212.99anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Merck Reference: 14,352 UN No.: 1438 MDL No.: MFCD00003419

Vzorec: Al(NO₃)₃·9H₂O
Hustota: 1,72 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149132
CAS číslo: 7784-27-2
EINECS: 236-751-8
UN: 1438
ADR: 5.1,III
Merck Index: 14,352

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR