Tisk…

Lanthanum(III) sulphate hydrate ≥99.9%

Dodavatel: Thermo Fisher Scientific

Synonyms: Dilanthanum trisulphate hydrate

43106.22 43106.36
43106.22EA 3050 CZK
43106.22 43106.36
Lanthanum(III) sulphate hydrate ≥99.9%
Lanthanum(III) sulphate hydrate

Purity: 99.9% CAS No.: 57804-25-8 Molecular Formula: La2(SO4)3.xH2O Formula Weight: 566.00(anhy) Physical Form: Crystalline Density: 2.821 Melting Point: dec. white heat Merck Reference: 14,5363 MDL No.: MFCD00011070

Vzorec: La₂O₁₂S₃·H₂O
Bod tání: Dec. (white heat)
Hustota: 2,821
Storage Temperature: Ambient
MDL Number: MFCD00149760
CAS číslo: 57804-25-8
EINECS: 233-239-6
Merck Index: 14,5363

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR