Tisk…

Ruthenium(IV) sulphide ≥99.9% (metals basis)

Dodavatel: Thermo Scientific
Varování

43098.06
43098.06EA 18770 CZK
43098.06
Ruthenium(IV) sulphide ≥99.9% (metals basis)
Ruthenium(IV) sulphide

Purity: 99.9% CAS No.: 12166-20-0 Molecular Formula: RuS2 Formula Weight: 165.19 Physical Form: Powder UN No.: UN3178 MDL No.: MFCD00799846

Vzorec: RuS₂
MW: 165,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00799846
CAS číslo: 12166-20-0
EINECS: 235-318-0
UN: 3178
ADR: 4.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR