Tisk…

Niob hard ≥99.8% (metals basis), rods, hard, Ø 19 mm (0.75 in)

Dodavatel: Thermo Fisher Scientific

43080.BS 43080.BP
43080.BSEA 12740 CZK
43080.BS 43080.BP
Niob hard ≥99.8% (metals basis), rods, hard, Ø 19 mm (0.75 in)
Niobium

Purity: 99.8% CAS No.: 7440-03-1 Physical Form: ≈24.3g/cm, Temper: as drawn Odour: Odourless MDL No.: MFCD00011126

Vzorec: Nb
Bod varu: 3700 °C (1013 hPa)
Bod tání: 2468 °C
Hustota: 8,57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS číslo: 7440-03-1
EINECS: 231-113-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR