Tisk…

Železo ≥99.995% (metals basis), wire, Puratronic®, Ø 0.075 mm (0.003 in)

Dodavatel: Thermo Fisher Scientific

42948.G6 42948.H2
42948.G6EA 5770 CZK
42948.G6 42948.H2
Železo ≥99.995% (metals basis), wire, Puratronic®, Ø 0.075 mm (0.003 in)
Železo

Purity: 99.995% CAS No.: 7439-89-6 Physical Form: ≈0.03g/m MDL No.: MFCD00010999

Vzorec: Fe
Bod varu: 3000 °C (1013 hPa)
Bod tání: 1535 °C
Hustota: 7,86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS číslo: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR