Tisk…

Oxid titanatý ≥99.9% (metals basis), pellets

Dodavatel: Thermo Fisher Scientific

42940.14 42940.22
42940.14EA 5770 CZK
42940.14 42940.22
Oxid titanatý ≥99.9% (metals basis), pellets
Oxid titanatý

Purity: 99.9% CAS No.: 12137-20-1 Molecular Formula: TiO Formula Weight: 63.90 Physical Form: 8-9mm(0.32-0.36in) dia x 6-7mm(0.24-0.28in) thick Density: 4.95 Melting Point: 1700° Boiling Point: 3000° MDL No.: MFCD00036279

Vzorec: OTi
Bod varu: >3000 °C
Bod tání: 1700 °C
Hustota: 4,95
Storage Temperature: Ambient
MDL Number: MFCD00036279
CAS číslo: 12137-20-1
EINECS: 235-236-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR