Tisk…

Calcium-3-hydroxy-3-methylbutyrate hydrate ≥97% (by titrimetric analysis), white powder

Dodavatel: Thermo Fisher Scientific
Varování

42721.18 42721.30 42721.A1
42721.18EA 3020 CZK
42721.18 42721.30 42721.A1
Calcium-3-hydroxy-3-methylbutyrate hydrate ≥97% (by titrimetric analysis), white powder
Calcium-3-hydroxy-3-methylbutyrate hydrate

Purity: 97+% CAS No.: 135236-72-5 Molecular Formula: C10H18CaO6.xH2O Formula Weight: 274.33(anhy) Physical Form: Powder Appearance: White MDL No.: MFCD01318562

Vzorec: C₁₀H₁₈CaO₆·H₂O
Storage Temperature: Ambient
MDL Number: MFCD01321171
CAS číslo: 135236-72-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR