Tisk…

Strontium titanate substrate 5x5x1 mm, polished one side 100 orientation

Dodavatel: Thermo Fisher Scientific

42621.KU
42621.KUEA 13700 CZK
42621.KU
Strontium titanate substrate 5x5x1 mm, polished one side 100 orientation
Strontium titanate

CAS No.: 12060-59-2 Molecular Formula: SrTiO3 Odour: Odourless MDL No.: MFCD00049554

Vzorec: O₃SrTi
Bod tání: 2080 °C
Hustota: 5,11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049554
CAS číslo: 12060-59-2
EINECS: 235-044-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR