Tisk…

Aluminium oxide, α-phase ≥99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific

Synonyms: ɑ-Tonerde, α-Alumina

42571.22 42571.36 42571.A3
42571.22EA 1090 CZK
42571.22 42571.36 42571.A3
Aluminium oxide, α-phase ≥99.9% (metals basis)
Aluminium oxide, α-phase

Purity: 99.9% CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 20-50 Micron APS, Spherical Powder, S.A. 5-6m2/g Refractive Index: 1.765 Odour: Odourless Melting Point: 2045° Boiling Point: 2980° Merck Reference: 14,356 MDL No.: MFCD00003424

Vzorec: Al₂O₃
Bod varu: 2980 °C
Bod tání: 2045 °C
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS číslo: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR