Tisk…

Potassium standard dissolved in oil, 5.000 µg/g K in mineral oil Specpure® pro wear metal analysis

Dodavatel: Thermo Fisher Scientific

42448.18
42448.18EA 3380 CZK
42448.18
Potassium standard dissolved in oil, 5.000 µg/g K in mineral oil Specpure® pro wear metal analysis
Standardy Standardy kationické Standardy - draslík

Physical Form: Liquid
MDL No.: MFCD00133776

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR