Tisk…

Molybden ≥99.95% (metals basis), slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)

Dodavatel: Thermo Fisher Scientific

42361.09 42361.18
42361.09EA 1540 CZK
42361.09 42361.18
Molybden ≥99.95% (metals basis), slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Molybden

Purity: 99.95% CAS No.: 7439-98-7 Physical Form: ≈0.26g/slug MDL No.: MFCD00003465

Vzorec: Mo
Bod varu: 3617 °C (1013 hPa)
Bod tání: 2622 °C
Hustota: 10,28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS číslo: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR