Tisk…

Vanad ≥99.8% (metals basis), wire, Ø 1.5 mm (0.06 in)

Dodavatel: Thermo Fisher Scientific

42224.BS 42224.BW 42224.CB
42224.BSEA 2170 CZK
42224.BS 42224.BW 42224.CB
Vanad ≥99.8% (metals basis), wire, Ø 1.5 mm (0.06 in)
Vanad

Purity: 99.8% CAS No.: 7440-62-2 Physical Form: ≈10.8g/m Odour: Odourless MDL No.: MFCD00011453

Vzorec: V
Bod varu: 3477 °C (1013 hPa)
Bod tání: 1896 °C
Hustota: 6,015 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011453
CAS číslo: 7440-62-2
EINECS: 231-171-1
Merck Index: 12,10053

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR