Tisk…

Grafit, plate, Thickness: 6.35 mm (0.250 in)

Dodavatel: Thermo Fisher Scientific

41831.GHEA 6760 CZK
41831.GH
Grafit, plate, Thickness: 6.35 mm (0.250 in)
Grafit
Vzorec: C
MW: 12.01
Bod tání: 3827
Hustota: 2.09…2.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS číslo: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR