Tisk…

Sodium diphenylamine-4-sulphonate ACS

Dodavatel: Thermo Fisher Scientific
Varování

41714.09 41714.04 41714.18
41714.09EA 4840 CZK
41714.09 41714.04 41714.18
Sodium diphenylamine-4-sulphonate ACS
Sodium diphenylamine-4-sulphonate

CAS No.: 6152-67-6 Molecular Formula: C12H10NNaO3S Formula Weight: 271.27 Physical Form: Powder MDL No.: MFCD00036468


Redox indicator

Vzorec: C₁₂H₁₀NNaO₃S
Storage Temperature: Ambient
MDL Number: MFCD00036468
CAS číslo: 6152-67-6
EINECS: 228-165-6
Merck Index: 13,07397

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR