Tisk…

4,4'-Diaminobiphenyl-2,2'-disulphonic acid hydrate, (∼30% H₂O)

Dodavatel: Thermo Fisher Scientific
Varování

41472.14 41472.22
41472.14EA 2050 CZK
41472.14 41472.22
4,4'-Diaminobiphenyl-2,2'-disulphonic acid hydrate, (∼30% H₂O)
4,4'-Diaminobiphenyl-2,2'-disulphonic acid hydrate

CAS No.: 117-61-3 Molecular Formula: C12H12N2O6S2.xH2O Formula Weight: 344.36(anhy) Physical Form: Powder Appearance: Dark purple MDL No.: MFCD00041885

Vzorec: C₁₂H₁₂N₂O₆S₂·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00041885
CAS číslo: 117-61-3
EINECS: 204-200-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR