Tisk…

trans-Bis(triphenylphosphine)palladium(II) chloride ≥99.95% (metals basis) Pd ≥ 14.7%, Premion®

Dodavatel: Thermo Fisher Scientific

41245.03 41245.06 41245.14
41245.03EA 2470 CZK
41245.03 41245.06 41245.14
trans-Bis(triphenylphosphine)palladium(II) chloride ≥99.95% (metals basis) Pd ≥ 14.7%, Premion®
trans-Bis(triphenylphosphine)palladium(II) chloride

Purity: 99.95% CAS No.: 13965-03-2 Molecular Formula: PdCl2[P(C6H5)3]2 Formula Weight: 701.91 Physical Form: Powder Sensitivity: Hygroscopic Appearance: Yellow Odour: Odourless Melting Point: ca 310° dec. MDL No.: MFCD00009593

Vzorec: C₃₆H₃₀Cl₂P₂Pd
Storage Temperature: Ambient
MDL Number: MFCD00009593
CAS číslo: 13965-03-2
EINECS: 237-744-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR