Tisk…

Di-sec-butoxyaluminoxytriethoxysilane

Dodavatel: Thermo Fisher Scientific
Varování

41210.09 41210.18 41210.30
41210.09EA 1710 CZK
41210.09 41210.18 41210.30
Di-sec-butoxyaluminoxytriethoxysilane
Di-sec-butoxyaluminoxytriethoxysilane

CAS No.: 68959-06-8 Molecular Formula: C14H33AlO6Si Formula Weight: 352.48 Flash Point: 46°(115°F) Physical Form: Liquid Density: 1.00 Sensitivity: Moisture Sensitive Appearance: Colourless Boiling Point: 200° UN No.: UN1993 MDL No.: MFCD00053771

Vzorec: C₁₄H₃₃AlO₆Si
Bod varu: >200 °C
Hustota: 1,00
Bod vzplanutí: 46 °C (115 °F)
Storage Temperature: Ambient
MDL Number: MFCD00053771
CAS číslo: 68959-06-8
EINECS: 273-392-6
UN: 1993
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR