Tisk…

Oxid niobičný ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), thickness 3.18 mm (0.125 in)

Dodavatel: Thermo Fisher Scientific

Synonyms: Diniobium pentaoxide, Niobium pentoxide

41080.KS
41080.KSEA 21790 CZK
41080.KS
Oxid niobičný ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), thickness 3.18 mm (0.125 in)
Oxid niobičný

Purity: 99.95% Molecular Formula: Nb2O5 Formula Weight: 265.81 MDL No.: MFCD00011128


Magnetron sputtering target

Vzorec: Nb₂O₅
Bod tání: 1496 °C
Hustota: 4,6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011128
CAS číslo: 1313-96-8
EINECS: 215-213-6
Merck Index: 14,06559

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR