Tisk…

Tris-(hydroxymethyl)aminomethan hydrochlorid ≥99.0% (dried basis), Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®

Dodavatel: Avantor
Varování

Synonyms: Tris(hydroxymethyl)methylamine hydrochloride, 2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride, Trizma® hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, TRIS HCl, Tromethane hydrochloride, Tris-HCl, Tris(hydroxymethyl)methylammonium chloride, Trometamol hydrochloride, THAM HCl

4103-07EA 97820 CZK
4103-07 4103-08 4103-01 4103-06 4103-02 4103-04 4103-F7
Tris-(hydroxymethyl)aminomethan hydrochlorid ≥99.0% (dried basis), Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
Tris-(hydroxymethyl)aminomethan hydrochlorid

For liquid chromatography and molecular biology applications.


  • Lot analysis on label

Vzorec: NH₂C(CH₂OH)₃·HCl
MW: 157.6
Bod tání: 150…152 °C
Hustota: 1.05 g/ml (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012590
CAS číslo: 1185-53-1
EINECS: 214-684-5
Merck Index: 12,09902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cbdc9446fdb9bcfe3f1d992ff20a16726f98113&vid=3d668801395b81fa595401a752279006da8fd3cb&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

For Laboratory,Research,or Manufacturing Use ≥99.0%
Assay (NH₂C(CH₂OH)₃HCl) (dried basis) ≥ 99.0 %
Appearance Passes Test
Absorbance of a 1M Solution - 260 nm ≤ 0.06
Absorbance of a 1M Solution - 280 nm ≤ 0.06
Absorbance of a 1M Solution - 400 nm ≤ 0.01
Water (H₂O)(by Karl Fischer titrn) ≤ 0.5 %
RNase Activity (None Detected) Passes Test
DNase Activity (None Detected) Passes Test
Protease Activity (None Detected) Passes Test
Trace Impurities - Calcium (Ca) ≤ 1 ppm
Trace Impurities - Copper (Cu) ≤ 1 ppm
Trace Impurities - Iron (Fe) ≤ 1 ppm
Trace Impurities - Lead (Pb) ≤ 1 ppm
Trace Impurities - Magnesium (Mg) ≤ 1.000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR