Tisk…

Železo ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)

Dodavatel: Thermo Fisher Scientific

40855.KS
40855.KSEA 19870 CZK
40855.KS
Železo ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Železo

Purity: 99.95% CAS No.: 7439-89-6 MDL No.: MFCD00010999

Vzorec: Fe
Bod varu: 3000 °C (1013 hPa)
Bod tání: 1535 °C
Hustota: 7,86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS číslo: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR