Tisk…

Zlato ≥99.95% (metals basis), wire, annealed, Ø 0.025 mm (0.001 in)

Dodavatel: Thermo Fisher Scientific

40585.G1 40585.G5 40585.G8
40585.G1EA 2750 CZK
40585.G1 40585.G5 40585.G8
Zlato ≥99.95% (metals basis), wire, annealed, Ø 0.025 mm (0.001 in)
Zlato

Purity: 99.95% CAS No.: 7440-57-5 Physical Form: ≈0.01g/m Odour: Odourless MDL No.: MFCD00003436

Vzorec: Au
Bod varu: 2800 °C (1013 hPa)
Bod tání: 1063 °C
Hustota: 19,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS číslo: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR