Tisk…

Bis(cyclopentadienyl)titanium dichloride ≥99%

Dodavatel: Thermo Fisher Scientific
Varování


Packaged under argon.

40386.04 40386.09 40386.18
40386.04EA 954 CZK
40386.04 40386.09 40386.18
Bis(cyclopentadienyl)titanium dichloride ≥99%
Bis(cyclopentadienyl)titanium dichloride

Purity: 99+% CAS No.: 1271-19-8 Molecular Formula: C10H10Cl2Ti Formula Weight: 249.00 Physical Form: Crystalline, Packaged under argon Density: 1.60 Appearance: Red Odour: Odourless Melting Point: ca 285° dec. Merck Reference: 14,9482 MDL No.: MFCD00003723

Vzorec: C₁₀H₁₀Cl₂Ti
Hustota: 1,60
Storage Temperature: Ambient
MDL Number: MFCD00003723
CAS číslo: 1271-19-8
EINECS: 215-035-9
UN: 3261
ADR: 8,II
Merck Index: 14,9482

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR