Tisk…

Hydrogenuhličitan draselný 99.7-100.5% (dried basis) ACS

Dodavatel: Thermo Fisher Scientific

40195.22 40195.36
40195.22EA 731 CZK
40195.22 40195.36
Hydrogenuhličitan draselný 99.7-100.5% (dried basis) ACS
Hydrogenuhličitan draselný

Purity: 99.7-100.5% CAS No.: 298-14-6 Molecular Formula: KHCO3 Formula Weight: 100.12 Physical Form: Granular Density: 2.17 Solubility: Slightly soluble in water. Practically insoluble in alcohol Merck Reference: 14,7609 MDL No.: MFCD00011402

Vzorec: KHCO₃
Bod tání: 292 °C
Hustota: 2,17
Storage Temperature: Ambient
MDL Number: MFCD00011402
CAS číslo: 298-14-6
EINECS: 206-059-0
Merck Index: 14,7609

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR