Tisk…

Oxid měďný ≥99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Cuprous oxide, Dicopper oxide

40189.06 40189.14 40189.22 40189.36
40189.06EA 2110 CZK
40189.06 40189.14 40189.22 40189.36
Oxid měďný ≥99.9% (metals basis)
Oxid měďný

Purity: 99.9% CAS No.: 1317-39-1 Molecular Formula: Cu2O Formula Weight: 143.08 Physical Form: Powder Density: 6.0 Refractive Index: 2.705 Sensitivity: Air and Moisture Sensitive Appearance: Red to dark red Odour: Odourless Melting Point: 1235° Boiling Point: 1800° dec. Merck Reference: 14,2664 UN No.: 3077 MDL No.: MFCD00010974

Vzorec: Cu₂O
Bod varu: 1800 °C (rozkl.)
Bod tání: 1235 °C
Hustota: 6,0
Storage Temperature: Ambient
MDL Number: MFCD00010974
CAS číslo: 1317-39-1
EINECS: 215-270-7
UN: 3077
ADR: 9,III
Merck Index: 14,2664

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR