Tisk…

Grafit, plate 1.27x9.98x9.98 mm (0.05x0.393x.393 in), pyrolytic coated

Dodavatel: Thermo Fisher Scientific

40156.KF
40156.KFEA 8720 CZK
40156.KF
Grafit, plate 1.27x9.98x9.98 mm (0.05x0.393x.393 in), pyrolytic coated
Grafit
Vzorec: C
MW: 12.01
Bod tání: 3827
Hustota: 2.09…2.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS číslo: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR