Tisk…

Anti-IL-4 Rat Monoclonal Antibody (In Vivo Ready™) [clone: MP4-25D2]

Dodavatel: Tonbo Biosciences
40-7048-U100 40-7048-U500
40-7048-U100EA 1900 CZK
40-7048-U100 40-7048-U500
Anti-IL-4 Rat Monoclonal Antibody (In Vivo Ready™) [clone: MP4-25D2]
Protilátky
The MP4-25D2 antibody is specific for human IL-4, a pleiotropic cytokine that Th2 cell differentiation, B cell proliferation and differentiation, and B cell class switching to IgE. MP4-25D2 is a neutralizing antibody and is reported to cross react with rhesus monkey IL-4.

Type: Primary
Antigen: IL-4
Clonality: Monoclonal
Clone: MP4-25D2
Conjugation: In Vivo Ready™
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Human, Rhesus
Objednat

Produktová specifikace

Symbol antigenu IL4
Název antigenu Interleukin 4
Environmentally Preferable
Synonyma antigenu BCGF-1|B-cell stimulatory factor 1|IL-4|Interleukin 4|BCGF1|BSF1|BSF-1|Interleukin-4
Druh protilátky Primární
Klonovalita Monoclonal
Klon MP4-25D2
Reaktivita Rhesus monkey,Human
Hostitel Rat
Identifikační číslo genu 3565
Izotyp IgG1 kappa
Průtoková cytometrie Yes
Skladovací teplota +2 °C to +8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR