Tisk…

Gold(III) acetate ≥99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific

39742.02 39742.03 39742.06
39742.02EA 5220 CZK
39742.02 39742.03 39742.06
Gold(III) acetate ≥99.9% (metals basis)
Gold(III) acetate

Purity: 99.9% CAS No.: 15804-32-7 Molecular Formula: Au(O2CCH3)3 Formula Weight: 374.10 Physical Form: Powder

Vzorec: C₆H₉AuO₆
Storage Temperature: Ambient
MDL Number: MFCD00210614
CAS číslo: 15804-32-7
EINECS: 244-137-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR