Tisk…

Hliník ≥99.9% (metals basis), foil, Thickness: 0.5 mm (0.02 in)

Dodavatel: Thermo Fisher Scientific

39733.FY 39733.GD
39733.FYEA 3540 CZK
39733.FY 39733.GD
Hliník ≥99.9% (metals basis), foil, Thickness: 0.5 mm (0.02 in)
Hliník

Purity: 99.9% CAS No.: 7429-90-5 Physical Form: ≈6.7g/50x100mm, Temper: as rolled Odour: Odourless MDL No.: MFCD00134029

Vzorec: Al
Bod varu: 2327 °C (1013 hPa)
Bod tání: 660 °C
Hustota: 2,7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS číslo: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR