Tisk…

Molybden ≥99.9% (metals basis), powder APS ∼2-4 µm

Dodavatel: Thermo Fisher Scientific
Varování

39687.14 39687.22
39687.14EA 2230 CZK
39687.14 39687.22
Molybden ≥99.9% (metals basis), powder APS ∼2-4 µm
Molybden

Purity: 99.9% CAS No.: 7439-98-7 Appearance: Grey Odour: Odourless UN No.: 3089 MDL No.: MFCD00003465

Vzorec: Mo
Bod varu: 5560 °C (1013 hPa)
Bod tání: 2622 °C
Hustota: 10,2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS číslo: 7439-98-7
EINECS: 231-107-2
UN: 3089
ADR: 4.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR