Tisk…

Lithium salicylate, tech.

Dodavatel: Thermo Fisher Scientific
Varování

39356.14 39356.22 39356.36
39356.14EA 1320 CZK
39356.14 39356.22 39356.36
Lithium salicylate, tech.
Lithium salicylate

CAS No.: 552-38-5 Molecular Formula: C7H5LiO3 Formula Weight: 144.05 Physical Form: Powder Odour: Sweet Melting Point: dec. MDL No.: MFCD00045813

Vzorec: C₇H₅LiO₃
Bod tání: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00045813
CAS číslo: 552-38-5
EINECS: 209-011-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR