Tisk…
Advanced TC™ cell culture flasks
Dodavatel: Greiner Bio-One
690975_200 658970_120 658975_120 660960_50 660975_50 661975_40
392-0259EA 14080 CZK
392-0259 392-0234 392-0235 392-0236 392-0237 392-0241
Advanced TC™ cell culture flasks
Baňky Baňky pro buněčné kultury
Cell culture flasks with a standard or ventilated cap and a modified culture surface to optimise cell characteristics and functions. The improved adherence and increased growth rate improve and accelerate cell proliferation. Additionally, the Advanced TC™ surface facilitates uniform and consistent cell adherence, increases throughput and reduces cell loss, for example during automated washing stages. Free of detectable DNase, RNase and human DNA. Free of cytotoxic substances.
  • Standard or ventilated cap
  • Increased cell adherence
  • Uniform and consistent cell
More Product Information